missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 887826
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent voltage-dependent NaV1.8 channel blocker (IC50 = 11 nM in recombinant human NaV1.8 channels). Reduces tactile allodynia in neuropathic rat models. 3- and 28-fold more potent at NaV1.8 channels compared to NaV1.2 and NaV1.7 channels respectively.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 480.99 |
| Purity | >97% |
| Molecular Weight (g/mol) | 480.993 |
| Chemical Name or Material | A 887826 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction