missing translation for 'onlineSavingsMsg'
Learn More

A 844606, TRC

Product Code. 30179567
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179567 10 mg 10mg
30178953 50 mg 50mg
30179215 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179567 Supplier Toronto Research Chemicals Supplier No. TRCA10123010MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 861119-08-6
Molecular Formula C20H20N2O2
Molecular Weight (g/mol) 320.38
Synonym A 844606, 2-(7-Methyl-3,7-diazabicyclo[3.3.0]octan-3-yl)xanthen-9-one, 2-(Hexahydro-5-methylpyrrolo[3,4-c]pyrrol-2(1H)-yl)-9H-xanthen-9-one
IUPAC Name 2-(5-methylhexahydropyrrolo[3,4-c]pyrrol-2(1H)-yl)-9H-xanthen-9-one
SMILES CN1CC2CN(CC2C1)c3ccc4Oc5ccccc5C(=O)c4c3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H20N2O2/c1-21-9-13-11-22(12-14(13)10-21)15-6-7-19-17(8-15)20(23)16-4-2-3-5-18(16)24-19/h2-8,13-14H,9-12H2,1H3
Formula Weight 320.15
Chemical Name or Material 2-(Hexahydro-5-methylpyrrolo[3,4-c]pyrrol-2(1H)-yl)-9H-xanthen-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.