missing translation for 'onlineSavingsMsg'
Learn More

A-836339, MedChemExpress

Product Code. 16445328
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16445328

Brand: Medchem Express HY12761

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-836339 is a cannabinoid CB2 receptor-selective agonist; exhibits high potencies at CB(2) and selectivity over CB(1) receptors.

A-836339 is a cannabinoid CB2 receptor-selective agonist

Specifications

Chemical Name or Material A-836339
Physical Form Solid
CAS 959746-77-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H26N2O2S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 12 mg/mL (38.65 mM; Need ultrasonic and warming)
SMILES CC1=C(C)S/C(N1CCOC)=N\C(C2C(C)(C)C2(C)C)=O
Molecular Weight (g/mol) 310.45
Formula Weight 310.45
Percent Purity 99.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.