missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies A 83-01
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
- A 83-01 is a potent inhibitor of activin receptor-like kinase (ALK) including ALK5 (type I transforming growth factor-β receptor), ALK4 (type IB activin receptor), and ALK7 (type I NODAL receptor) with IC50 = 12, 45, and 7.5 nM, respectively (Tojo et al.)
- Facilitates the conversion of epiblast stem cells to the naїve pluripotent state (Zhou et al.; Li et al.)
- In combination with PD0325901, enables OCT4-mediated reprogramming (Zhu et al.)
- Inhibits mesoderm and endoderm specification (Zhang et al.)
- Disrupts epithelial to mesenchymal transition through inhibition of the SMAD signaling pathway (Tojo et al.)
Specifications
Specifications
| CAS | 909910-43-6 |
| Quantity | 10 mg |
| Molecular Formula | C25H19N5S |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction