missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 804598
GREENER_CHOICE

Product Code. 18095102
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18095102

Brand: Tocris Bioscience™ 4473/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective P2X7 antagonist

Potent, competitive P2X7 receptor antagonist (IC50 values are 8.9, 9.9 and 10.9 nM for mouse, rat and human P2X7 receptors respectively). Selective for P2X7 receptors (IC50 > 5-10 μM for a wide array of cell surface receptors and ion channels). Binds with high affinity (Ki app = 2.4 nM for rat P2X7 receptors).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125758-85-1
Molecular Formula C19H17N5

Specifications

Quantity 10 mg
Formula Weight 315.37
Purity >99%
Molecular Weight (g/mol) 315.38
Chemical Name or Material A 804598
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.