missing translation for 'onlineSavingsMsg'
Learn More

A-802715, MedChemExpress

Product Code. p-200095925
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16483179

Brand: Medchem Express HYU00142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A802715 is a methylxanthine derivative. A802715 has a TD50 (toxic dose of 50%) of 0.9-1.1 mM.

A802715 is a methylxanthine derivative. A802715 has a TD50 (toxic dose of 50%) of 0.9-1.1 mM.

Specifications

Chemical Name or Material A-802715
CAS 107767-58-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H26N4O3
Quantity 1 mg
Solubility Information DMSO : 190 mg/mL (589.33 mM; Need ultrasonic)
SMILES O=C(N1CCCCC(C)(O)C)N(C)C2=C(N(CCC)C=N2)C1=O
Molecular Weight (g/mol) 322.4
Formula Weight 322.4
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.