missing translation for 'onlineSavingsMsg'
Learn More

A-802715, MedChemExpress

Product Code. 16483179
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483179 1 mg 1mg
16442398 5 mg 5mg
16440199 10 mg 10mg
16430716 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483179 Supplier Medchem Express Supplier No. HYU00142

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress A802715 is a methylxanthine derivative. A802715 has a TD50 (toxic dose of 50%) of 0.9-1.1 mM.

A802715 is a methylxanthine derivative. A802715 has a TD50 (toxic dose of 50%) of 0.9-1.1 mM.

Specifications

Chemical Name or Material A-802715
CAS 107767-58-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H26N4O3
Quantity 1 mg
Solubility Information DMSO : 190 mg/mL (589.33 mM; Need ultrasonic)
SMILES O=C(N1CCCCC(C)(O)C)N(C)C2=C(N(CCC)C=N2)C1=O
Molecular Weight (g/mol) 322.4
Formula Weight 322.4
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.