missing translation for 'onlineSavingsMsg'
Learn More

A-770041, MedChemExpress

Product Code. 16454358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454358 10 mg 10mg
16417077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16454358 Supplier Medchem Express Supplier No. HY11011
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress A-770041 is selective and orally active Src-family Lck inhibitor; A-770041 is a 147 nM inhibitor of Lck (1 mM ATP) and is 300-fold selective against Fyn, the other Src family kinase involved in T-cell signaling.

A-770041 is selective and orally active Src-family Lck inhibitor

Specifications

Chemical Name or Material A-770041
CAS 869748-10-7
Color White
Purity Grade Notes Research
Molecular Formula C34H39N9O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (160.84 mM)
SMILES NC1=C2C(N([C@@H]3CC[C@@H](N4CCN(C(C)=O)CC4)CC3)N=C2C5=CC=C(NC(C6=CC7=C(C=CC=C7)N6C)=O)C(OC)=C5)=NC=N1
Molecular Weight (g/mol) 621.73
Formula Weight 621.73
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.