missing translation for 'onlineSavingsMsg'
Learn More

A-769662, MedChemExpress

Product Code. 16450538
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450538

Brand: Medchem Express HY50662

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM.

A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM.

Specifications

Chemical Name or Material A-769662
CAS 844499-71-4
Color Light Brown
Purity Grade Notes Research
Molecular Formula C20H12N2O3S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (138.74 mM; ultrasonic and warming and heat to 80°C)
SMILES N#CC1C(C2=C(NC1=O)SC=C2C3=CC=C(C4=CC=CC=C4O)C=C3)=O
Molecular Weight (g/mol) 360.39
Formula Weight 360.39
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.