missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 740003

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A 740003 is a potent and selective P2X7 receptor antagonist (IC50 values are 18 and 40 nM for rat and human receptors respectively). Displays selectivity over a variety of P2X and P2Y receptors up to a concentration of 100 μM. Reduces nociception in animal models of persistent neuropathic and inflammatory pain. Also reduces neuroblastoma tumor growth in mice.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | A 740003 |
| Purity | >98% |
| Molecular Weight (g/mol) | 474.55 |
| Inhibitors | A 740003 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction