missing translation for 'onlineSavingsMsg'
Learn More

A-674563 hydrochloride, MedChemExpress

Product Code. 16495588
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495588

Brand: Medchem Express HY13254A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-674563 hydrochloride is a potent and selective Akt1 inhibitor with Ki of 11 nM.

A-674563 hydrochloride is a potent and selective Akt1 inhibitor with Ki of 11 nM.

Specifications

Chemical Name or Material A-674563 hydrochloride
Color Light Brown
Purity Grade Notes Research
Molecular Formula C22H23ClN4O
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (253.23 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (158.27 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=CC=C1)COC2=CC(C3=CC4=C(C=C3)NN=C4C)=CN=C2.[H]Cl
Molecular Weight (g/mol) 394.9
Formula Weight 394.9
Percent Purity 99.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.