missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ A 61603 hydrobromide

Product Code. 30001407
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001407 10 mg 10mg
18025462 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001407 Supplier R&D Systems™ Supplier No. 1052/10
  • ONLINE EXCLUSIVE PRICE
    £208.00
    £196.35 / 10mg
    Save £11.65
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α1A agonist

Potent α-adrenoceptor agonist that is at least 35-fold more potent at α1A than at α1B or α1D sites. Induces dose response increases in spontaneous Ca2+ transients in rat ventricular myocytes in vitro (EC50 = 6.9 nmol/L).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107756-30-9
Molecular Formula C14H20BrN3O3S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 390.29
Purity >98%
Molecular Weight (g/mol) 390.296
Chemical Name or Material A 61603 hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.