missing translation for 'onlineSavingsMsg'
Learn More

A-549 (human lung carcinoma) Nuclear extract lysate, Non-denatured; Abnova

Product Code. 16049575
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 16049575

Brand: Abnova L025V4.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Western Blotting, Immunoprecipitation

Specifications

For Use With (Application) Immunoprecipitation, Western Blot
Tissue Lung
Description Nuclear extract cell lysate (non-denatured)
Lysis Buffer Buffer A: 10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT. Buffer C: 20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. Buffer D : 20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF.
Preparation Method Nuclear extract was prepared by using a modified protocol of Dignam et al. Cells were Harvested and homogenized in Buffer A, and then centrifugated at 25,000 g for 20 minutes to remove cytoplasm and pellet the nuclei. The pellet was re-suspended in Buffer C, and then the suspensions were centrifuged to collect nuclear extract. The supernatant was dialyzed against Buffer D. The dialysate was then centrifuged, divided into aliquots, and stored at -80°C. The protein concentration was determined by the method of Bradford (Bio-Rad protein assay, microplate standard assay). The lysate was adjusted to 2 mg/ml.
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 50 μg
Storage Buffer In Buffer D.
Host Species Human
Concentration 2.5 mg/mL
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.