missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 485

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
A 485 is a potent and selective p300/CREB-binding protein (CBP) HAT domain inhibitor (IC50 values are 2.6 and 9.8 nM for the CBP-bromodomain HAT-C/H3 (BHC) and p300-BHC domains, respectively), which displays > 1000-fold selectivity over closely related HATs. A 485 suppresses proliferation in several hematological malignancies and AR+ prostate cancer cell lines in vitro, and also inhibits tumor growth in a castration-resistant prostate cancer xenograft model. A 485 is orally bioavailable.
To request the negative control for A 485, please fill out the A 486 request form on the SGC website.
Specifications
Specifications
| CAS | 1889279-16-6 |
| Synonym | (1R)-N-[(4-Fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide |
| Quantity | 25 mg |
| Molecular Formula | C25H24F4N4O5 |
| Formula Weight | 536.48 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction