missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 485
GREENER_CHOICE

Product Code. 18733018
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18733018

Brand: Tocris Bioscience™ 6387/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Histone Acetyltransferase Inhibitors

A 485 is a potent and selective p300/CREB-binding protein (CBP) HAT domain inhibitor (IC50 values are 2.6 and 9.8 nM for the CBP-bromodomain HAT-C/H3 (BHC) and p300-BHC domains, respectively), which displays > 1000-fold selectivity over closely related HATs. A 485 suppresses proliferation in several hematological malignancies and AR+ prostate cancer cell lines in vitro, and also inhibits tumor growth in a castration-resistant prostate cancer xenograft model. A 485 is orally bioavailable.

To request the negative control for A 485, please fill out the A 486 request form on the SGC website.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1889279-16-6
Synonym (1R)-N-[(4-Fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide
Quantity 5 mg
Molecular Formula C25H24F4N4O5
Formula Weight 536.48
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.