missing translation for 'onlineSavingsMsg'
Learn More

AZD-3463, MedChemExpress

Product Code. 16416688
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416688

Brand: Medchem Express HY15609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

Specifications

Chemical Name or Material AZD-3463
CAS 1356962-20-3
Color Green
Purity Grade Notes Research
Molecular Formula C24H25ClN6O
Quantity 10 mg
Synonym ALK/IGF1R inhibitor
Solubility Information DMSO : 20 mg/mL (44.55 mM; Need ultrasonic)
SMILES ClC(C=NC(NC1=C(OC)C=C(N2CCC(N)CC2)C=C1)=N3)=C3C4=CNC5=C4C=CC=C5
Molecular Weight (g/mol) 448.95
Formula Weight 448.95
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.