missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-431 (human epidermoid carcinoma) Whole cell lysate, Denatured; Abnova
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Description
Western Blotting
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Epidermis |
| Description | Whole cell lysate (denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was mixed in 5X Sample Buffer to become final 1X Sample Buffer as Storage Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 200 μg |
| Storage Buffer | In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). |
| Host Species | Human |
| Concentration | 3 mg/mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction