missing translation for 'onlineSavingsMsg'
Learn More

A 419259 trihydrochloride, MedChemExpress

Product Code. 16472877
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472877

Brand: Medchem Express HY15764A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A 419259 trihydrochloride is a Src family kinases inhibitor with IC50s of 9 nM, 3 nM and 3 nM for Src, Lck and Lyn, respectively.

A 419259 trihydrochloride is a Src family kinases inhibitor with IC50s of 9 nM, 3 nM and 3 nM for Src, Lck and Lyn, respectively.

Specifications

Chemical Name or Material A 419259 trihydrochloride
CAS 1435934-25-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H37Cl3N6O
Quantity 50 mg
Synonym RK 20449 trihydrochloride
Solubility Information H2O : ≥ 55 mg/mL (92.91 mM) ∣DMSO : 1 mg/mL (1.69 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3CC[C@H](N4CCN(C)CC4)CC3)C=C2C5=CC=C(OC6=CC=CC=C6)C=C5)=NC=N1.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 592.0
Formula Weight 592.0
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.