missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-385358, MedChemExpress
Description
A-385358 is a selective inhibitor of Bcl-XL with Kis of 0.80 and 67 nM for Bcl-XL and Bcl-2, respectively.
Specifications
Specifications
| Chemical Name or Material | A-385358 |
| CAS | 406228-55-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C32H41N5O5S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (78.15 mM; Need ultrasonic) |
| SMILES | O=C(NS(=O)(C1=CC=C(N[C@@H](CSC2=CC=CC=C2)CCN(C)C)C([N+]([O-])=O)=C1)=O)C3=CC=C(N4CCC(C)(C)CC4)C=C3 |
| Molecular Weight (g/mol) | 639.83 |
| Formula Weight | 639.83 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?