missing translation for 'onlineSavingsMsg'
Learn More

A-381393, MedChemExpress

Product Code. 16471117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471117 50 mg 50mg
16443266 25 mg 25mg
16434868 10 mg 10mg
16427617 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471117 Supplier Medchem Express Supplier No. HY116941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-381393 is a potent, selective, brain penetrate dopamine D4 receptor antagonist, with Kis of 1.5, 1.9 and 1.6 nM for human dopamine D4.4, D4.2, and D4.7 receptor, respectively, >2700-fold selectivity over D1, D2, D3 and D5 dopamine receptors. A-381393 shows moderate affinity for 5-HT2A (Ki, 370 nM).

A-381393 is a potent, selective, brain penetrate dopamine D4 receptor antagonist, with Kis of 1.5, 1.9 and 1.6 nM for human dopamine D4.4, D4.2, and D4.7 receptor, respectively, >2700-fold selectivity over D1, D2, D3 and D5 dopamine receptors. A-381393 shows moderate affinity for 5-HT2A (Ki, 370 nM).

Specifications

Chemical Name or Material A-381393
Physical Form Solid
CAS 726174-00-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H24N4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 51.67 mg/mL (161.25 mM; Need ultrasonic)
SMILES CC1=CC=C(C=C1C)N2CCN(CC2)CC3=NC4=CC=CC=C4N3
Molecular Weight (g/mol) 320.43
Formula Weight 320.43
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.