missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 366

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A 366 is a potent and selective G9a/GLP histone lysine methyltransferase inhibitor (IC50 = 3.3 nM). Exhibits >1000-fold selectivity for G9a/GLP over 21 other methyltransferases. Decreases levels of lysine 9 dimethylation on histone H3 (H3K9Me2) in PC3 cells.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | A 366 |
| Purity | >98% |
| Molecular Weight (g/mol) | 329.44 |
| Inhibitors | A 366 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction