missing translation for 'onlineSavingsMsg'
Learn More

A-317491 sodium salt hydrate, MedChemExpress

Product Code. 16489197
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489197 5 mg 5mg
16462727 50 mg 50mg
16446638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489197 Supplier Medchem Express Supplier No. HY15568A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress A-317491 sodium salt hydrate is a potent, selective and non-nucleotide antagonist of P2X3 and P2X2/3 receptors, with Kis of 22, 22, 9, and 92 nM for hP2X3, rP2X3, hP2X2/3, and rP2X2/3, respectively.

A-317491 sodium salt hydrate is highly selective (IC50>10 μM) over other P2 receptors and other neurotransmitter receptors, ion channels, and enzymes. A-317491 sodium salt hydrate reduces inflammatory and neuropathic pain by blocking P2X3 and P2X2/3 receptor-mediated calcium flux.

Specifications

Chemical Name or Material A-317491 sodium salt hydrate
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H29NNaO9
Quantity 5 mg
Solubility Information H2O : ≥ 100 mg/mL (164.86 mM) ∣DMSO : ≥ 100 mg/mL (164.86 mM)
SMILES O=C(C1=CC(C(N(CC2=CC=CC(OC3=CC=CC=C3)=C2)[C@H]4CCCC5=C4C=CC=C5)=O)=C(C(O)=O)C=C1C(O)=O)O.[Na].O
Molecular Weight (g/mol) 606.57
Formula Weight 606.57
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.