missing translation for 'onlineSavingsMsg'
Learn More

A-317491, MedChemExpress

Product Code. 16479197
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479197 5 mg 5mg
16452727 50 mg 50mg
16436638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16479197 Supplier Medchem Express Supplier No. HY15568
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress A-317491 is a potent, selective and non-nucleotide antagonist of P2X3 and P2X2/3 receptors, with Kis of 22, 22, 9, and 92 nM for hP2X3, rP2X3, hP2X2/3, and rP2X2/3, respectively. A-317491 is highly selective (IC50>10 μM) over other P2 receptors and other neurotransmitter receptors, ion channels, and enzymes. A-317491 reduces inflammatory and neuropathic pain by blocking P2X3 and P2X2/3 receptor-mediated calcium flux.

A-317491 is a potent, selective and non-nucleotide antagonist of P2X3 and P2X2/3 receptors, with Kis of 22, 22, 9, and 92 nM for hP2X3, rP2X3, hP2X2/3, and rP2X2/3, respectively. A-317491 is highly selective (IC50>10 μM) over other P2 receptors and other neurotransmitter receptors, ion channels, and enzymes. A-317491 reduces inflammatory and neuropathic pain by blocking P2X3 and P2X2/3 receptor-mediated calcium flux.

Specifications

Chemical Name or Material A-317491
CAS 475205-49-3
Color Pink
Purity Grade Notes Research
Molecular Formula C33H27NO8
Quantity 5 mg
Solubility Information DMSO : ≥ 47 mg/mL (83.10 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC(C(N(CC2=CC=CC(OC3=CC=CC=C3)=C2)[C@H]4CCCC5=C4C=CC=C5)=O)=C(C(O)=O)C=C1C(O)=O)O
Molecular Weight (g/mol) 565.57
Formula Weight 565.57
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.