missing translation for 'onlineSavingsMsg'
Learn More

a-(3,4-Diethoxyphenyl)-6,7-diethoxy-1-isoquinolinemethanol, TRC

Product Code. 30381196
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381196 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381196 Supplier Toronto Research Chemicals Supplier No. TRCD44213510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-(3,4-Diethoxyphenyl)-6,7-diethoxy-1-isoquinolinemethanol
CAS 1394077-64-5
Molecular Formula C24H29NO5
Quantity 10 mg
Synonym (6,7-Diethoxyisoquinolin-1-yl)(3,4-diethoxyphenyl)methanol
InChI Formula InChI=1S/C24H29NO5/c1-5-27-19-10-9-17(14-20(19)28-6-2)24(26)23-18-15-22(30-8-4)21(29-7-3)13-16(18)11-12-25-23/h9-15,24,26H,5-8H2,1-4H3
SMILES CCOC1=C(OCC)C=C(C=CN=C2C(C3=CC=C(OCC)C(OCC)=C3)O)C2=C1
Molecular Weight (g/mol) 411.49
Formula Weight 411.2046
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.