missing translation for 'onlineSavingsMsg'
Learn More

A-1210477, MedChemExpress

Product Code. 16497907
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497907 5 mg 5mg
16495876 100 mg 100mg
16445188 10 mg 10mg
16441407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497907 Supplier Medchem Express Supplier No. HY12468

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-1210477 is a potent and selective inhibitor of MCL-1 with a Ki of 0.45 nM. A-1210477 specifically binds MCL-1 and promotes apoptosis of cancer cells in an MCL-1-dependent manner.

A-1210477 is a potent and selective inhibitor of MCL-1 with a Ki of 0.45 nM. A-1210477 specifically binds MCL-1 and promotes apoptosis of cancer cells in an MCL-1-dependent manner.

Specifications

Chemical Name or Material A-1210477
CAS 1668553-26-1
Color White
Purity Grade Notes Research
Molecular Formula C46H55N7O7S
Quantity 5 mg
Solubility Information DMSO : 10 mg/mL (11.76 mM; Need ultrasonic)
SMILES O=C(C(N1CCN2CCOCC2)=C(CCCOC3=C4C=CC=CC4=CC=C3)C5=C1C(C6=C(COC7=CC=C(N8CCN(S(=O)(N(C)C)=O)CC8)C=C7)N(C)N=C6C)=CC=C5)O
Molecular Weight (g/mol) 850.04
Formula Weight 850.04
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.