missing translation for 'onlineSavingsMsg'
Learn More

A-1155463, MedChemExpress

Product Code. p-200101273
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480328 5 mg 5mg
16447868 10 mg 10mg
16423817 50 mg 50mg
16417686 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480328 Supplier Medchem Express Supplier No. HY19725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell.

A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell.

Specifications

Chemical Name or Material A-1155463
CAS 1235034-55-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H32FN5O4S2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (74.65 mM; Need ultrasonic)
SMILES O=C(C1=C(CCCOC2=CC=C(C#CCN(C)C)C=C2F)SC(N3CC4=C(C=CC=C4C(NC5=NC6=CC=CC=C6S5)=O)CC3)=N1)O
Molecular Weight (g/mol) 669.79
Formula Weight 669.79
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.