missing translation for 'onlineSavingsMsg'
Learn More

Dyphylline, Thermo Scientific™

An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator

Brand:  Thermo Scientific Alfa Aesar J63167.22

Quantity 100g

Product Code. 15465429

  • £75.90 / 100g
 Restricted Item
Estimated Shipment: 22-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

479-18-5
MFCD00005759
284563
dyphylline, diprophylline, diphylline, 7-2,3-dihydroxypropyl theophylline, diprophyllin, glyphylline, neothylline, lufyllin, aristophyllin, diprofilline
KSCFJBIXMNOVSH-UHFFFAOYSA-N
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
3182
254.24
C10H14N4O4
100g
14,3479
Soluble in water
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
254.246
CHEBI:4728
Dyphylline
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.