missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid sodium salt, 98%, Thermo Scientific™

NMDA receptor antagonist at the glycine site

Brand:  Thermo Scientific Alfa Aesar J61283.MA

Quantity 10mg

Product Code. 15453899

Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
7-Chlorokynurenic acid sodium salt is used as a Potent competitive inhibitor of L-glutamate transport into synaptic vesicles. It is also used as an NMDA receptor antagonist and inhibitor of EAAT.

Solubility
Soluble in water at 100mM

Notes
Store at room temperature (desiccating conditions). May be stored at 4°C. For long-term storage, aliquot and store at 4°C. Do not freeze. For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1263094-00-3
MFCD12195840
7-chlorokynurenic acid sodium salt
IFZYIORLNGNLEI-UHFFFAOYSA-M
sodium;2-carboxy-7-chloroquinolin-4-olate
124219644
≥99%
C10H5ClNNaO3
10mg
Soluble in water at 100mM
C1=CC2=C(C=C1Cl)N=C(C=C2[O-])C(=O)O.[Na+]
245.594
245.6
7-Chlorokynurenic acid sodium salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.