missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide-d{6}, 99.9% (Isotopic), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 036517.48

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15441457

  • £43.60 / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.18 g/mL
Characteristic
Hygroscopic
2206-27-1
C2H6OS
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
(²H₃)methanesulfinyl(²H₃)methane
75151
99.9%
88°C (190°F)
1.476
10 x 1g
99.9% (Isotopic)
MFCD00002090
Miscible with water and diethyl ether.
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
84.17
84.17
Dimethyl sulfoxide-d6, Isotopic
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.