missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60203.5H

63.84 GBP valid until 2026-09-25
BEST PRICE on Promo! Use promo code "27966" to get your promotional price.


Quantity 454g

Product Code. 15423019

  • ONLINE EXCLUSIVE PRICE
    £67.20
    £60.65 / 454g
    Save £6.55
    9% Off
View all promotions
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

Solubility
Soluble in ethanol (moderately), DMSO, alkali hydroxides, ammonia, dilute hyrochloric acid, nitric acid, dilute aqueous acid, and dilute aqueous base. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

58-55-9
C7H8N4O2
454g
13463
theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur
ZFXYFBGIUFBOJW-UHFFFAOYSA-N
1,3-dimethyl-7H-purine-2,6-dione
2153
180.16
Odorless
MFCD00079619
UN2811
14,9285
Soluble in ethanol (moderately),DMSO,alkali hydroxides,ammonia,dilute hyrochloric acid,nitric acid,dilute aqueous acid,and dilute aqueous base. Insoluble in water.
CN1C2=C(C(=O)N(C1=O)C)NC=N2
180.167
CHEBI:28177
Theophylline
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-09-25
Curiosity Goes Beyond
Promo Code: 27966
Push the boundaries of discovery with exceptional savings across essential research workflows: Cell Culture and Analysis driven by our Gibco™ product portfolio, Gene Expression powered by Applied Biosystems™ solutions, and Protein Expression enabled by Invitrogen™ and Thermo Scientific™ products.

Applying Promo and Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.