Learn More
Galanthamine hydrobromide, Thermo Scientific™
Centrally active and long-acting acetylcholinesterase inhibitor
Brand: Thermo Scientific Alfa Aesar J62482.MF
| Quantity | 50mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1953-04-4 | |
| MFCD00067672 | |
| UN2811 | |
| galanthamine hydrobromide, galantamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide | |
| QORVDGQLPPAFRS-XPSHAMGMSA-N | |
| 368.27 | |
| 368.27 |
| C17H22BrNO3 | |
| 50mg | |
| 14,4340 | |
| Soluble in water or DMSO | |
| CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br | |
| 121587 | |
| Galanthamine hydrobromide |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.