missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid, 50 mg/ml in water, sterile-filtered, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J67194.XF

Product Code. 15408669

  • £59.00 / 1mL
 Restricted Item
Estimated Shipment: 22-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Nalidixic acid is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. It is used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent)t DNA supercoiling, and chromosome fragmentation.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (0.9 mg/ml) at 23°C, methanol (1.3 mg/ml) at 23°C, toluene (1.6 mg/ml) at 23°C, DMSO (≥ 6 mg/ml), and solution of carbonates. Fully miscible in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

389-08-2
MFCD00006884
nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
4421
Nalidixic acid, 50mg/mL in water, sterile-filtered
C12H12N2O3
1mL
Soluble in chloroform (50mg/ml),ethanol (0.9mg/ml) at 23°C,methanol (1.3mg/ml) at 23°C,toluene (1.6mg/ml) at 23°C,DMSO (? 6mg/ml),and solution of carbonates. Fully miscible in water.
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
232.239
CHEBI:100147
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.