Learn More
Gilson™ Service Kit for Gilson Pipetman™
Brand:  Gilson™ FA07002
Product Code. 12908310
  • ONLINE EXCLUSIVE PRICE
    £49.70
    £47.79 / Pack of 1
    Save £1.91
    3% Off
Only null left
In Stock
Add to basket
Specifications
1st Level service kit
Pipetman™ P20G
Tip holder, 3 piston seals guide and 3 O-rings
1 Each