missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 042577.49

Product Code. 11379188

  • £603.00 / 100g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20667-12-3
99.99% (metals basis)
MFCD00003404
231.74
Light sensitive
7.2 g/cm3 at 20°C
4 x 25g
Ag2O
Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol
Odorless
Dark brown powder
Silver(I) oxide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.