Learn More
Kartell™ Polypropylene Watch Glasses
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ Polypropylene Watch Glass has good heat and chemical resistance.
Brand:  Kartell™ 0045600
Product Code. 10448842
  • £29.45 / Pack of 10
Only null left
Estimated Shipment: 20-02-2026
Add to basket
Description
Description
  • Very stable
  • Moulded base
  • Can withstand temperatures up to 120°C