missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopurine, 98%, Thermo Scientific™
Click to view available options
Glass bottle; 50mg
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 452-06-2 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 280.0°C to 284.0°C |
Color | Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min |
Packaging | Glass bottle |
Molecular Formula | C5H5N5 |
MDL Number | MFCD00005566 |
Show More |
Safety and Handling
Product Identifier
- 2-Aminopurine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'