missing translation for 'onlineSavingsMsg'
Learn More

9H-Fluorene-9-acetic acid, 99%, ACROS Organics™

Product Code. 10401163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10401163 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10401163 Supplier Thermo Scientific Acros Supplier No. 330710010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6284-80-6
CAS Min % 98.5
CAS Max % 100.0
Melting Point 131°C to 135°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C15H12O2
MDL Number MFCD00013262
Quantity 1 g
Synonym 9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid
InChI Key WFSMJMTYIMFHPV-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O
IUPAC Name 2-(9H-fluoren-9-yl)acetic acid
Molecular Weight (g/mol) 224.259
PubChem CID 94852
Formula Weight 224.26
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 9H-Fluorene-9-acetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.