missing translation for 'onlineSavingsMsg'
Learn More

9H-Carbazol-3-ol, TRC

Product Code. 30357743
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357743 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357743 Supplier Toronto Research Chemicals Supplier No. TRCC175815250MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9H-Carbazol-3-ol
CAS 7384-07-8
Molecular Formula C12H9O
Quantity 250 mg
Synonym 3-Hydroxycarbazole
InChI Formula InChI=1S/C13H11NO/c1-8-6-12-10(7-13(8)15)9-4-2-3-5-11(9)14-12/h2-7,14-15H,1H3
SMILES Oc1ccc2[nH]c3ccccc3c2c1
IUPAC Name 2-methyl-9H-carbazol-3-ol
Molecular Weight (g/mol) 183.21
Formula Weight 183.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.