missing translation for 'onlineSavingsMsg'
Learn More

(9E)-12-Hydroxy-9-octadecenoic Acid, TRC

Product Code. 30391720
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391720 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391720 Supplier Toronto Research Chemicals Supplier No. TRCH2967855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (9E)-12-Hydroxy-9-octadecenoic Acid
CAS 82188-83-8
Molecular Formula C18 H34 O3
Quantity 5 mg
Synonym (9E)-12-Hydroxy-9-octadecenoic Acid, 9-Octadecenoic acid, 12-hydroxy-, (9E)-, 9-Octadecenoic acid, 12-hydroxy-, (E)-, (9E)-12-Hydroxy-9-octadecenoic acid
InChI Formula InChI=1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9+
SMILES CCCCCCC(O)C\C=C\CCCCCCCC(=O)O
IUPAC Name (E)-12-hydroxyoctadec-9-enoic acid
Molecular Weight (g/mol) 298.461
Formula Weight 298.251
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.