missing translation for 'onlineSavingsMsg'
Learn More

9Beta,11Beta-Epoxy-17,21-dihydroxy-16Beta-methylpregna-1,4-diene-3,20-dione 21-Acetate, TRC

Product Code. 30008809
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008809 1 g 1g
17963065 10 mg 10mg
17951465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008809 Supplier Toronto Research Chemicals Supplier No. TRCE5891851G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912-38-9
Molecular Formula C24H30O6
Molecular Weight (g/mol) 414.49
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9,11-epoxy-17-hydroxy-16-methyl-, (9β,11β,16β)-, 9β-Pregna-1,4-diene-3,20-dione, 9,11β-epoxy-17,21-dihydroxy-16β-methyl-, 21-acetate (6CI,7CI,8CI), (9β,11β,16β)-21-(Acetyloxy)-9,11-epoxy-17-hydroxy-16-methylpregna-1,4-diene-3,20-dione, 9,11β-Epoxy-17-hydroxy-16β-methyl-3,20-dioxo-9β-pregna-1,4-diene 21-acetate, 9,11β-Epoxy-17,21-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione 21-acetate, 9,11β-Epoxy-17-hydroxy-16β-methyl-3,20-dioxo-9β-pregna-1,4-dien-21-yl Acetate
IUPAC Name [2-[(1S,2S,10S,11S,13S,14R,15S,17S)-14-hydroxy-2,13,15-trimethyl-5-oxo-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadeca-3,6-dien-14-yl]-2-oxoethyl] acetate
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H30O6/c1-13-9-18-17-6-5-15-10-16(26)7-8-21(15,3)24(17)20(30-24)11-22(18,4)23(13,28)19(27)12-29-14(2)25/h7-8,10,13,17-18,20,28H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,20-,21-,22-,23-,24+/m0/s1
Formula Weight 414.2042
Chemical Name or Material 9beta,11beta-Epoxy-17,21-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 21-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.