missing translation for 'onlineSavingsMsg'
Learn More

9alpha-Hydroxy 1,2-Dihydro Betamethasone 17,21-Dipropionate, TRC

Product Code. 30393992
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393992 2.5 mg 2.5mg
30391602 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393992 Supplier Toronto Research Chemicals Supplier No. TRCH8251802.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9a-Hydroxy 1,2-Dihydro Betamethasone 17,21-Dipropionate
Analyte or Component Names 9a-Hydroxy 1,2-Dihydro Betamethasone 17,21-Dipropionate
Shipping Condition Room Temperature
Molecular Formula C28H40O8
Synonym (9α,11β,16β)-9,11-Dihydroxy-16-methyl-17,21-bis(1-oxopropoxy)-pregna-4-ene-3,20-dione
InChI Formula InChI=1S/C28H40O8/c1-6-23(32)35-15-22(31)28(36-24(33)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-25(17,4)27(19,34)21(30)14-26(20,28)5/h13,16,19-21,30,34H,6-12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28?/m0/s1
Solution Type Neat
SMILES O[C@H]1C[C@@]2(C)[C@](C[C@H](C)C2(OC(CC)=O)C(COC(CC)=O)=O)([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@]31O)=O
IUPAC Name (8S,9R,10S,11S,13S,14S,16S)-9,11-dihydroxy-10,13,16-trimethyl-3-oxo-17-(2-(propionyloxy)acetyl)-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl propionate
Industry Type Forensics
Molecular Weight (g/mol) 504.61
Quantity 2.5 mg
Formula Weight 504.27
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.