missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate |
| Analyte or Component Names | 9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H35 F O7 |
| Synonym | 6-desfluoro Difluprednate |
| InChI Formula | InChI=1S/C27H35FO7/c1-5-6-23(33)35-26(22(32)15-34-16(2)29)12-10-19-20-8-7-17-13-18(30)9-11-24(17,3)27(20,28)21(31)14-25(19,26)4/h9,11,13,19-21,31H,5-8,10,12,14-15H2,1-4H3/t19-,20-,21-,24-,25-,26+,27-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)COC(=O)C |
| IUPAC Name | [(8S,9R,10S,11S,13S,14S,17S)-17-(2-acetoxyacetyl)-9-fluoro-11-hydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] butanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?