missing translation for 'onlineSavingsMsg'
Learn More

9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate, TRC

Product Code. 30394715
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394715 5 mg 5mg
30397192 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394715 Supplier Toronto Research Chemicals Supplier No. TRCF5958405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate
Analyte or Component Names 9Alpha-Fluoro Prednisolone 17-Butyrate-21-acetate
Shipping Condition Room Temperature
Molecular Formula C27 H35 F O7
Synonym 6-desfluoro Difluprednate
InChI Formula InChI=1S/C27H35FO7/c1-5-6-23(33)35-26(22(32)15-34-16(2)29)12-10-19-20-8-7-17-13-18(30)9-11-24(17,3)27(20,28)21(31)14-25(19,26)4/h9,11,13,19-21,31H,5-8,10,12,14-15H2,1-4H3/t19-,20-,21-,24-,25-,26+,27-/m0/s1
Solution Type Neat
SMILES CCCC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)COC(=O)C
IUPAC Name [(8S,9R,10S,11S,13S,14S,17S)-17-(2-acetoxyacetyl)-9-fluoro-11-hydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] butanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 490.56
Quantity 5 mg
Formula Weight 490.2367
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.