Learn More
Eppendorf™ 96-Well U-Bottom PCR Microplates

Pipette, control and recover samples with this polypropylene microplate. Eppendorf™ 96-Well U-Bottom PCR Microplates are ideal for assays that require high resistance against temperature or solvents.
Brand: Eppendorf™ 0030609204
Description
- Unmatched, transparent polypropylene (PP) for improved sample visibility
- OptiTrack™ Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax™ well design: Optimized well geometry for minimal remaining volume and excellent mixing properties
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- g-Safe™: exceptional centrifugation stability up to 6,000 × g
- Polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers or biocides – Materials which could have a negative effect on bioassays
- Guaranteed functionality from -86 to +100°C
- Autoclavable (121°C, 20 min.)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.