Learn More
Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates
GREENER_CHOICE
Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.
Brand:  Eppendorf™ 0030128729
Product Code. 10052423
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.
  • Improved well-to-well tolerance
  • Semi-skirt for labelling
  • Extremely thin-walled for optimum heat transfer into the sample
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing and reduced cross-contamination risk
  • 250μL well volume (when used with cap strips)