missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Septa for 96-Well Plates, for 3500/SeqStudio™ Flex
Shop All Applied Biosystems Products
Click to view available options
Quantity:
20 each
Unit Size:
Pack of 20
Specifications
Specifications
| Content And Storage | One bag of 20 septa. Store at room temperature. |
| Format | 96-well Plate |
| For Use With (Equipment) | 3500 Genetic Analyzer, 3500xL Genetic Analyzer |
| Product Type | Septa Plate |
| Quantity | 20 each |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction