Learn More
Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips
Product Code. p-7075504
Click to view available options
Surface Treatment:
Microlon 200
Microlon 600
Unit Size:
Pack of 100
Product Code. 10778242
Brand: Greiner Bio-One 754070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips are suited for agglutination tests.
  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Pipetted simply and cleanly
  • Suitable for +/- analyses
  • Suitable for transmission measurements
  • U-bottom (round bottom)
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 6
Volume (Metric) Working 50 to 280 μL
Sterility Non-sterile
Surface Treatment Microlon 200
Total Volume (Metric) Well 312 μL
Well Shape Round Bottom (U-well)
Dnase Free Yes
No. per Case 100
No. per Pack 5
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear