Learn More
Porvair Sciences™ 96-Well Polystyrene Microplates
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Material | Polystyrene |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 350 μL |