missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips

Product Code. 10770175
Change view
Click to view available options
Surface Treatment:
FLUOTRAC 600
LUMITRAC 600
Medium Binding
Microlon 600
For Use With (Application):
Fluorescence measurements
Luminescence measurements
Transmission measurements
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment For Use With (Application) Unit Size
10770175 LUMITRAC 600 Luminescence measurements Pack of 100
10105111 Medium Binding Transmission measurements Pack of 100
10073221 Microlon 600 Transmission measurements Pack of 100
10630973 Medium Binding Luminescence measurements Pack of 100
10080282 Medium Binding Fluorescence measurements Pack of 100
10274322 FLUOTRAC 600 Fluorescence measurements Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10770175 Supplier Greiner Bio-One Supplier No. 762074
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips are suitable for optical measurements.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Flat Bottom
  • Polystyrene
TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Format Strip Plate
Array 8 x 12
Binding Type High Binding
Color White
Product Type ELISA Strip Plate
Material Polystyrene
Volume (Metric) Well 388 μL
No. of Wells 96
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment LUMITRAC 600
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Luminescence measurements
No. per Case 100
No. per Pack 5
RNase Free Yes
Skirt Color White
Temperature Range (Metric) -20°C to +60°C
Well Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.