missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips
GREENER_CHOICE

Product Code. 10080282
Click to view available options
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
Microlon 200
Microlon 600
For Use With (Application):
Fluorescence measurements
Luminescence measurements
Transmission measurements
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10080282

Brand: Greiner Bio-One 762076

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips are suitable for optical measurements.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Flat Bottom
  • Polystyrene
TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Array 8 x 12
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment FLUOTRAC 200
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Fluorescence measurements
No. per Case 100
No. per Pack 5
RNase Free Yes
Skirt Color White
Temperature Range (Metric) -20°C to +60°C
Well Color Black
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.